Mouse recombinant IL-3
Artikelnr. ENZOALX2018210010
Lieferant: ENZO LIFE SCIENCES
CAS-Nummer:
Spezifikationen
- VE:10 µG
- Protein/Peptid-Typ:Rekombinant
- Spezies:Mouse
- Seq:02
- Biologische Aktivität:The activity is determined by the dose-dependent stimulation of TF-1 cells which is typically observed at concentations <0,1 ng/ml.
- Proteinsynonym:MCGF|HCGF|Multi-CSF|P-cell stimulation factor|Interleukin-3
- Protein/Peptid-Name:IL-3
- Purity:>97% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Molecular weight:~15,1kDa
- Zusammensetzung:Lyophilized. Sterile filtered.
Spezifikationen
Über diesen Artikel
Produced in E. coli. Non-glycosylated protein, containing 135 amino acids.
- Carrier-free