ContactUsLinkComponent
Human recombinant CREB binding protein (catalytic domain) (from E. coli)
  ENZOBMLSE4520100
 :  
Human recombinant CREB binding protein (catalytic domain) (from E. coli)
  ENZOBMLSE4520100
 :  
  • VE:
    100 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Seq:
    02
  • Protein/Peptid-Name:
    CREB binding protein (catalytic domain)
  • Purity:
    >65% (SDS-PAGE)
  • Zusammensetzung:
    Liquid. In 50 mM TRIS-HCl, pH 8,0, containing 0,1 mM EDTA, 10% glycerol.
Produced in E. coli. Catalytic domain (aa 1319-1710) of human CBP.