Human recombinant PKCepsilon (GST-tag),GST tag
ENZOBMLSE4430005
:
- VE:5 µG
- Protein/Peptid-Typ:Rekombinant
- Spezies:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/Peptid-Name:PKCepsilon (GST-tag)
Produced in insect cells. Full length active PKCε (protein kinase C ε).