Express Eingabe
Austria
AuftragsverfolgungKontakt
Z-RR-pNA

Z-RR-pNA

25 mg
Catalog # ENZOBMLP1380025
Supplier:   Enzo Life Sciences
Z-RR-pNA
Catalog # ENZOBMLP1380025
Supplier:  Enzo Life Sciences
CAS Number:  
Z-RR-pNA

25 mg

CAS Number:  
Price
170,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Cathepsin substrate

Chromogenic substrate specific for cathepsin B. Release of pNA in monitored at 405 to 410 nm. This substrate is useful for inhibitor screening and kinetic analysis. Also available: calibration standard pNA (BML-KI145).

Specifications

Specifications

  • VE:
    25 mg
  • Cat. No.:
    ENZOBMLP1380025
  • Purity:
    99%
  • Protein/Peptid-Name:
    Z-RR-pNA

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.