Order Entry
Austria
ContactUsLinkComponent
Human Recombinant EDA-A1 (soluble) (from E. coli)
Artikelnr. ENZOALX522038C010
Lieferant:  ENZO LIFE SCIENCES
CAS-Nummer:  
Human Recombinant EDA-A1 (soluble) (from E. coli)
Artikelnr. ENZOALX522038C010
Lieferant:  ENZO LIFE SCIENCES
CAS-Nummer:  

Spezifikationen

  • VE:
    10 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
  • Protein/Peptid-Name:
    EDA-A1 (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Konzentration:
    0,1 mg/ml after reconstitution
  • Zusammensetzung:
    Lyophilised. Contains PBS.

Spezifikationen

Über diesen Artikel

Produced in E. coli. The extracellular domain of human EDA-A1 (ectodysplasin-A1) (aa 245-391) is fused at the N-terminus to a linker peptide (9 aa) and a FLAG®-tag.