Ac-IETD-AFC
Caspase and granzyme substrate
Fluorogenic substrate for caspase-8/caspase-3 processing enzyme and granzyme B. Also a substrate for caspase-10. The IETD sequence is based on caspase-3 proenzyme cleavage site.
- Protein/Peptid-Name:Ac-IETD-AFC
- Purity:>98% (HPLC)
Frequently Bought Together