Reagents for fluoride determination using Lovibond® MD photometers
Catalog # LOVI467481
Supplier: Lovibond
CAS Number:
Specifications
- VE:250 mL
- Bezeichnung:SPADNS reagent solution
- Seq:44
- Für Gerät:MD 100, MD 600, MD610, MultiDirect and SpectroDirect
- Messbereich:0,2 – 2 mg/l F