Non-ionic surfactant, Tergitol™ NP-9, Sigma-Aldrich®
Lieferant: Merck
Zertifikate
Über diesen Artikel
Tergitol™ is a nonionic surfactant which can be used to extract estrogens.
Tergitol™ has been used in a study to assess a method to determine nickel using ultrasonic-assisted cloud point extraction. It has also been used in a study to investigate a method for the detection of Salmonella enteritidis via an enteritidis-specific real time PCR assay.
Turbidity: Clear to slightly hazy
Colour: Colourless to yellow
Cloud point: 52 to 56 °C
Spezifikationen
- Catalog No:
- SIALNP9-100ML
- SIALNP9-500ML
- M-Clarity Qualitätsstufe:
- MQ200
- MQ200