Organic reference standards, HMMNI (nitroimidazole metabolite NMIA P1296)
PROONMIAP1296
:
- VE:50 mg
- Normen:Reference standard
- CAS Nr.:936-05-0
- Bezeichnung:HMMNI
- Formel:C₅H₇N₃O₃
- Bezeichnung:HMMNI
Purity estimate obtained is 99,8 ± 1,3% mass fraction, (95% coverage interval).
- Pale yellow crystals
- Moisture content ≤0,1% mass fraction
- Volatile residue ≤0,1% mass fraction
- Non volatile residue ≤0,2% mass fraction
- CAS no.: 936-05-0
Synonyms: 2-hydroxymethyl-1-methyl-5-nitro-imidazole; 1-methyl-5-nitro-imidazole-2-methanol; 1-methyl-5-nitro-2-imidazole methanol