Standard reference material, strawberry leaves - trace elements
Catalog # PROOLGC7162
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- VE:20 g
- Bezeichnung:Strawberry leaves - trace elements
Specifications
About this item
This standard is intended for use in development, validation and quality control of analytical methods for the determination of elements in vegetation.