AuftragsverfolgungKontakt
Entacapone, Reference Standards
Supplier:   USP
Entacapone, Reference Standards
Supplier:  USP
Certificates
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
 : US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1235933.
  • 1235944.
  • Bezeichnung:
  • Entacapone
  • Entacapone Related Compound A
  • Synonym(s):
  • (Z)-2-cyano-3-(3,4-dihydroxy-5-nitrophenyl)-N,N-diethylacrylamide
  • CAS Nr.:
  • 130929-57-6
  • 145195-63-7
  • Formel:
  • C₁₄H₁₅N₃O₅
  • C₁₄H₁₅N₃O₅
Specifications
Product Family Options
Product Information
  • Bezeichnung
    Normen
    Packung
    VE
    Availability
    Price
  • Entacapone
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 674,00 €
    674,00 €
    1 * 200 mg
       674,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1235933.
    Bezeichnung
    Entacapone
    Synonym(s)
    CAS Nr.
    130929-57-6
    Formel
    C₁₄H₁₅N₃O₅
  • Entacapone Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 150,00 €
    1 150,00 €
    1 * 25 mg
       1 150,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1235944.
    Bezeichnung
    Entacapone Related Compound A
    Synonym(s)
    (Z)-2-cyano-3-(3,4-dihydroxy-5-nitrophenyl)-N,N-diethylacrylamide
    CAS Nr.
    145195-63-7
    Formel
    C₁₄H₁₅N₃O₅