PTFE coated GL 45 high purity silicone septum, DURAN®
Catalog # SCOT292483005
Supplier: DWK Life Sciences
Specifications
- Bezeichnung:PTFE coated GL 45 silicone septum for piercing
- VE:10
Specifications
About this item
A high purity PTFE coated silicone septum for use with DURAN® GL 45 open top screw caps and all sises of DURAN® GL 45 laboratory bottles. Can be used for sampling using a syringe and a needle.
- Soft silicone rubber for easy needle penetration and resealability
- Manufactured from high purity, Platinum catalysed silicone rubber
- High quality PTFE film reduces the risk of solvent contamination
- Wide temperature usage range from – 50 up to + 180°C
- Suitable for autoclaving and dry heat sterilisation
Applications: Liquid transfer from HPLC solvent bottles, aseptic reagent delivery or culture sampling using a syringe and a needle, reservoir cap for supplying lab instruments.