Human recombinant IFN gamma (from cells)
Artikelnr. PRSI92-362
Lieferant: ProSci Inc.
CAS-Nummer:
Spezifikationen
- VE:50 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:Cells
- Spezies:Human
- Lagerbedingungen:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Gen-ID:3458
- Proteinsynonym:Interferon Gamma|IFN-Gamma|IFNG|Immune Interferon
- Protein/Peptid-Name:IFN gamma
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:16.8 kD
- Sequence:Gln24-Gln166
- Zusammensetzung:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Validierte Anwendungen:Bioassay
Spezifikationen
Über diesen Artikel
IFN gamma is the major interferon produced by mitogenically or antigenically stimulated lymphocytes. It is structurally different from type I interferon and its major activity is immunoregulation. It has been implicated in the expression of class II histocompatibility antigens in cells that do not normally produce them, leading to autoimmune disease. Interferon gamma is produced mainly byT-cells and natural killer cells activated by antigens, mitogens, or alloantigens. It is produced by lymphocytes expressing the surface antigens CD4 and CD8. IFN gamma synthesis is induced by IL-2, FGF-basic, and EGF.
This recombinant protein can be used for biological assays. For research use only.