Order Entry
Austria
ContactUsLinkComponent
 
Human recombinant IL1 beta (from E. coli)
  PRSI92-219
 :  
Human recombinant IL1 beta (from E. coli)
  PRSI92-219
 :  

 

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Lagerbedingungen:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Proteinsynonym:
    Catabolin|IL-1b|IL1B.|Interleukin-1 beta|IL-1F2|IL1F2
  • Protein/Peptid-Name:
    IL1 beta
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Ala117-Ser269
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Validierte Anwendungen:
    Bioassay

 

 

IL1B belongs to the IL-1 family. Interleukin 1 (IL-1) is a family of polypeptide cytokines consisting of two agonists, IL-1 alpha (IL-1F1) and IL-1 beta (IL-1F2) encoded by two distinct genes and perform identical biological functions. IL-1 stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. IL-1 proteins are involved in the inflammatory response. It is identified as endogenous pyrogens, and is reported to stimulate the release of prostaglandin and collagenase from synovial cells.

This recombinant protein can be used for biological assays. For research use only.