Order Entry
Austria
ContactUsLinkComponent
Human recombinant siglec 9 (from cells)
Human recombinant siglec 9 (from cells)
Catalog # PRSI91-630
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant siglec 9 (from cells)
Catalog # PRSI91-630
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    Cells
  • Spezies:
    Human
  • Tag sequence:
    C-Fc tag
  • Lagerbedingungen:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Proteinsynonym:
    SIGLEC9|Siglec-9|CDw329|Protein FOAP-9|CD329|Sialic acid-binding Ig-like lectin 9
  • Protein/Peptid-Name:
    Siglec 9
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    63.2 kD
  • Sequence:
    Gln18-Gly348
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Validierte Anwendungen:
    Bioassay

Specifications

About this item

Sialic acid-binding Ig-like lectin 9(Siglec 9) is expressed by peripheral blood leukocytes (neutrophils and monocytes but not eosinophils), and found in liver, fetal liver, bone marrow, placenta, spleen and in lower levels in skeletal muscle, fetal brain and so on. It is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It also binds to alpha-2,3- or alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-Fc tag