Order Entry
Austria
ContactUsLinkComponent
Mouse recombinant Growth hormone (from E. coli)
Mouse recombinant Growth hormone (from E. coli)
Catalog # PRSI91-038
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant Growth hormone (from E. coli)
Catalog # PRSI91-038
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Mouse
  • Lagerbedingungen:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Proteinsynonym:
    Somatotropin|Growth Hormone|Gh|Gh1
  • Protein/Peptid-Name:
    Growth Hormone
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21.9 kD
  • Sequence:
    Phe27-Phe216
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of 50mM TrisHCl, 500mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Validierte Anwendungen:
    Bioassay

Specifications

About this item

Somatotropin(GH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. GH stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues.

  • This recombinant protein can be used for biological assays. For research use only.