Order Entry
Austria
ContactUsLinkComponent
Human recombinant siglec5 (from HEK293 cells)
Human recombinant siglec5 (from HEK293 cells)
Catalog # PRSI96-691
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant siglec5 (from HEK293 cells)
Catalog # PRSI96-691
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VE:
    100 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    HEK293 cells
  • Spezies:
    Human
  • Proteinsynonym:
    CD170|SIGLEC5|OB-BP2|CD33L2
  • Protein/Peptid-Name:
    Siglec5
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    73 kDa
  • Zusammensetzung:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Validierte Anwendungen:
    Western Blot

Specifications

About this item

Sialic acid-binding Ig-like lectin 5 (SIGLEC5) is also known as CD33 antigen-like 2 (CD33L2), Obesity-binding protein 2 (OB-BP2), CD antigen CD170, which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. SIGLEC5 / CD170 is expressed by monocytic / myeloid lineage cells. SIGLEC-5 / CD33-L2 binds equally to alpha-2,3-linked and alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.