Order Entry
Austria
ContactUsLinkComponent
Human recombinant BTLA (from HEK293 cells)
Human recombinant BTLA (from HEK293 cells)
Catalog # PRSI96-067
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant BTLA (from HEK293 cells)
Catalog # PRSI96-067
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VE:
    100 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    HEK293 cells
  • Spezies:
    Human
  • Proteinsynonym:
    BTLA|CD272
  • Protein/Peptid-Name:
    BTLA
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    38.7 kDa
  • Zusammensetzung:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Validierte Anwendungen:
    Western Blot

Specifications

About this item

B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.