Order Entry
Austria
ContactUsLinkComponent
 
Human recombinant PPP1CC (from E. coli)
  PRSIXW-RP3205
 :  
undefined
Human recombinant PPP1CC (from E. coli)
  PRSIXW-RP3205
 :  

 

  • VE:
    20 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Proteinsynonym:
    PP1C|PP-1G|PPP1GProtein phosphatase 1C catalytic subunit
  • Protein/Peptid-Name:
    PPP1CC
  • Purity:
    >95%
  • Molecular weight:
    37.1 kDa (Calculated)
  • Validierte Anwendungen:
    ELISA, Western Blot, MS

 

 

Protein phosphatase 1 (PP1) is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. Involved in regulation of ionic conductances and long-term synaptic plasticity. May play an important role in dephosphorylating substrates such as the postsynaptic density-associated Ca(2+)/calmodulin dependent protein kinase II.

  • This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.002% NaN, 3mM NaCl, 2.5mM

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.