Human ZIP14 peptide
PRSI6105P
:
- VE:50 µG
- Spezies:Human
- Lagerbedingungen:Store at –20 °C, stable for one year.
- Aminosäurezahl:16 amino acid peptide near the center of human ZIP14
- Protein/Peptid-Name:ZIP14 Peptide
- Validierte Anwendungen:Blocking
ZIP14 peptide is used for blocking the activity of ZIP14 antibody.