ContactUsLinkComponent
Human ZIP14 peptide
  PRSI6105P
 :  
Human ZIP14 peptide
  PRSI6105P
 :  
  • VE:
    50 µG
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at –20 °C, stable for one year.
  • Aminosäurezahl:
    16 amino acid peptide near the center of human ZIP14
  • Protein/Peptid-Name:
    ZIP14 Peptide
  • Validierte Anwendungen:
    Blocking
ZIP14 peptide is used for blocking the activity of ZIP14 antibody.