Anti-ADIPOQ Rabbit Polyclonal Antibody (Biotin)
Artikelnr. PRSI36-192
Lieferant: ProSci Inc.
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:Adiponectin
- Antigen-Symbol:ADIPOQ
- Klonalität:Polyklonal
- Konjugation:Biotin
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Form:Liquid
- Gen-ID:9370
- Antigen-Synonyme:APM1|APM-1|GBP28|Adipocyte Complement Related Protein of 30kDa|ADPN|ACDC|ACRP30|ADIPQTL1
- Lagerungspuffer:0.2um-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Lagertemperatur:Stable for at least 6 months after receipt when stored at -20˚C.
- Konzentration:1 mg/ml
- Transporttemperatur:-20°C
- Immunogen:Recombinant human adiponectin.
- Größe:50 μg
- VE:50 µG
Spezifikationen
Über diesen Artikel
ACRP30 was identified as a novel adipocyte-specific synthesized and secreted protein with structural resemblance to complement factor C1q. Like adipsin, ACRP30 secretion is induced ~10-fold during adipocyte differentiation. Plasma levels are reduced in obese humans, and low levels are associated with insulin-resistance. Treatment of db/db mice with TZD increased ACRP30 levels.
Type: Primary
Antigen: ADIPOQ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human