Express Eingabe
Austria
AuftragsverfolgungKontakt
Anti-EIF4E Rabbit Polyclonal Antibody | Avantor

Anti-EIF4E Rabbit Polyclonal Antibody

100 µg
Artikelnr. PRSI79-374
Anbieter:   ProSci Inc.
Anti-EIF4E Rabbit Polyclonal Antibody
Artikelnr. PRSI79-374
Anbieter:  ProSci Inc.
Anti-EIF4E Rabbit Polyclonal Antibody
Anti-EIF4E Rabbit Polyclonal Antibody

100 µg

Preis
411,00 €
Einloggen für Verfügbarkeit
1 * 100 µG
Product Details & Documents

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.

Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100, Immunofluorescence: 1:100~1:200

Antibodies were purified by affinity-chromatography using epitope-specific peptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Dokumente

Spezifikationen
  • Größe:
    100 µg
  • Western blot:
    Yes
  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Reaktivität:
    Human|Rat|Mouse
  • Konzentration:
    1 mg/mL
  • Cat. No.:
    PRSI79-374
  • ImmunoChemie:
    Yes
  • ImmunFluoreszenz:
    Yes
  • Immunogen:
    eIF4E (Ab-209) antibody was raised against a peptide sequence around aa. 207~211 (S-G-S-T-T) derived from Human eIF4E.
  • Antigen-Synonyme:
    eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eukaryotic translation initiation factor 4E-like 1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
  • Antigen-Symbol:
    EIF4E
  • Klonalität:
    Polyklonal
  • Wirt:
    Rabbit
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.