Specifications
- Antigen-Name:Crumbs Family Member 2
- Antigen-Symbol:CRB2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen-Synonyme:CRB2|Protein crumbs homolog 2|Crumbs-like protein 2
- Konzentration:1 mg/mL
- Immunogen:Rabbit polyclonal CRB2 antibody was raised against a 17 amino acid peptide near the center of human CRB2.
- Größe:100 µg
- VE:100 µG
Specifications
About this item
CRB2 Antibody: CRB2 (Crumbs homolog 2), like its homologs CRB1 and CRB3, is similar to the Drosophila crumbs protein and is expressed in retina, brain and kidney. Along with other proteins, the Crumbs proteins form a complex that help set up cell polarity in developing neuroepithelial cells. At the onset of neural specification, embryonic stem cells (ESCs) upregulate CRB2, which then localizes apically in neural rosettes. Gain- and loss-of-function studies of CRB2 have shown that CRB2 is essential for the stabilization of other polarity proteins. Unlike CRB1, mutations in CRB2 do not appear to play a role in retinitis pigmentosa or in Leber congenital amaurosis.
CRB2 Antibody is affinity chromatography purified via peptide column.
CRB2 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: