Order Entry
Austria
ContactUsLinkComponent
Anti-CLIC4 Goat Polyclonal Antibody
Anti-CLIC4 Goat Polyclonal Antibody
Catalog # PRSI46-645
Supplier:  ProSci Inc.
undefined
Anti-CLIC4 Goat Polyclonal Antibody
Catalog # PRSI46-645
Supplier:  ProSci Inc.

Specifications

  • Antigen-Name:
    Chloride Intracellular Channel 4
  • Antigen-Symbol:
    CLIC4
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Goat
  • ImmunFluoreszenz:
    Yes
  • Reaktivität:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen-Synonyme:
    H1|huH1|p64H1|CLIC4L|MTCLIC
  • Konzentration:
    500 ug/mL
  • Immunogen:
    CLIC4 antibody was raised against a 13 amino acid synthetic peptide near the N-Terminus of CLIC4.
  • Größe:
    100 µg
  • VE:
    100 µG

Specifications

About this item

ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 30 kDa band observed in human kidney and human placenta lysates (calculated MW of 28.8 kDa according to NP_039234.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Edwards et al, Am J Physiol. 1999 Mar;276 (3 Pt 2) :F398-408.; PMID: 10070163). Recommended concentration: 0.1-0.3 μg/mL.

CLIC4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CLIC4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CLIC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: