Anti-LAMA5 Goat Polyclonal Antibody
Artikelnr. PRSI42-418
Lieferant: ProSci Inc.
Spezifikationen
- Antigen-Name:Laminin, Alpha 5
- Antigen-Symbol:LAMA5
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Goat
- Western blot:Yes
- Form:Liquid
- Konzentration:500 ug/mL
- Immunogen:LAMA5 antibody was raised against a 15 amino acid synthetic peptide near the internal region of LAMA5 (aa 1029-1043).
- Größe:100 µg
- VE:100 µG
Spezifikationen
Über diesen Artikel
LAMA5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
LAMA5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LAMA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: