- Antigen-Name:Inhibitor Of Growth Family Member 1
- Antigen-Symbol:ING1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:p47|p33ING1b|p33ING1|p24ING1c|p33|p47ING1a
- Immunogen:A synthetic peptide corresponding a region of human ING1.
- Größe:100 µg
- VE:100 µG
ING1 is a tumor suppressor protein that can induce cell growth arrest and apoptosis. The protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of ING1 gene have been detected in various cancers.This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ING1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat