Anti-ZIK1 Rabbit Polyclonal Antibody
PRSI30-610
- Antigen-Name:Zinc Finger Protein Interacting with K Protein 1
- Antigen-Symbol:ZIK1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ZIK1.
- Größe:100 µg
- VE:100 µG
ZIK1 may be a transcriptional repressor.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZIK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human