Specifications
- Antigen-Name:Annexin A8 like2
- Antigen-Symbol:ANXA8L2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:ANXA8L2
- Immunogen:A synthetic peptide corresponding a region of human ANXA8L2.
- Größe:50 µg
- VE:50 µG
Specifications
About this item
ANXA8L2 is a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. ANXA8L2 may function as an an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of ANXA8L2 has been associated with acute myelocytic leukemia.This gene encodes a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. The encoded protein may function as an an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of this gene has been associated with acute myelocytic leukemia. A highly similar duplicated copy of this gene is found in close proximity on the long arm of chromosome 10.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ANXA8L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human