Anti-HELB Rabbit Polyclonal Antibody
Catalog # PRSI28-506
Supplier: ProSci Inc.
Specifications
- Antigen-Symbol:HELB
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human HELB.
- Größe:100 µg
- VE:100 µG
Specifications
About this item
The protein encoded by this gene is a DNA helicase. A dominant-negative mutant of this protein blocks chromosomal DNA replication and suggests that its function is required for S phase entry.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HELB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human