Anti-CTBP1 Rabbit Polyclonal Antibody
Artikelnr. PRSI27-608
Lieferant: ProSci Inc.
Spezifikationen
- Antigen-Name:C-Terminal Binding Protein 1
- Antigen-Symbol:CTBP1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:CTBP1|BARS|CtBP1|MGC104684|CTBP
- Immunogen:A synthetic peptide corresponding a region of human CTBP1.
- Größe:50 µg
- VE:50 µG
Spezifikationen
Über diesen Artikel
The phosphoprotein C-terminal binding protein 1 (CTBP-1) binds the C-terminus of adenovirus E1A protein. CTBP-1 is a transcriptional repressor and may play a role during cellular proliferation. A second closely related gene, CTBP2, maps to chromosome 21.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CTBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: