Anti-LEF1 Rabbit Polyclonal Antibody
Catalog # PRSI27-586
Supplier: ProSci Inc.
Specifications
- Antigen-Name:Lymphoid Enhancer-Binding Factor 1
- Antigen-Symbol:LEF1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Zebrafish,Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:TCF1ALPHA|TCF1-alpha|DKFZp586H0919|LEF-1|LEF1
- Immunogen:A synthetic peptide corresponding a region of human LEF1.
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Lymphoid enhancer-binding factor-1 (LEF1) is a 48-kD nuclear protein that is expressed in pre-B and T cells. It binds to a functionally important site in the T-cell receptor-alpha enhancer and confers maximal enhancer activity. LEF1 belongs to a family of regulatory proteins that share homology with high mobility group protein-1.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LEF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish