- Antigen-Name:Heat Shock 70kDa Protein 2
- Antigen-Symbol:HSPA2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:HSP70-3|HSP70-2
- Immunogen:A synthetic peptide corresponding a region of human HSPA2.
- Größe:50 µg
- VE:50 µG
HSPA2 belongs to the heat shock protein 70 family.In cooperation with other chaperones, HSPA2 stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HSPA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse