Anti-TSPYL4 Rabbit Polyclonal Antibody
Catalog # PRSI27-007
Supplier: ProSci Inc.
Specifications
- Antigen-Name:TSPY-like 4
- Antigen-Symbol:TSPYL4
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:D10Bwg0791e|B230210I21Rik|2610102M01Rik
- Immunogen:A synthetic peptide corresponding a region of human TSPYL4.
- Größe:50 µg
- VE:50 µG
Specifications
About this item
TSPYL4 belongs to the nucleosome assembly protein (NAP) family. The functions of TSPYL4 remain unknown.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TSPYL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat