Order Entry
Austria
ContactUsLinkComponent
Anti-FAM98A Rabbit Polyclonal Antibody
Artikelnr. PRSI26-988
Lieferant:  ProSci Inc.
undefined
Anti-FAM98A Rabbit Polyclonal Antibody
Artikelnr. PRSI26-988
Lieferant:  ProSci Inc.

Spezifikationen

  • Antigen-Name:
    Family With Sequence Similarity 98, Member A
  • Antigen-Symbol:
    FAM98A
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Rabbit
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen-Synonyme:
    FAM 98A|FA98A_HUMAN.|LOC25940|Protein FAM98A|Family with sequence similarity 98 member A|Hypothetical protein LOC25940
  • Immunogen:
    A synthetic peptide corresponding a region of human FAM98A.
  • Größe:
    50 µg
  • VE:
    50 µG

Spezifikationen

Über diesen Artikel

FAM98A belongs to the FAM98 family. The function of FAM98A remains unknown.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FAM98A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat