Specifications
- Antigen-Name:SLIT And NTRK-like Family member 1
- Antigen-Symbol:SLITRK1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:SLIT and NTRK-like protein 1|TTM|LRRC12|KIAA1910|Slitrk1|Leucine-rich repeat-containing protein 12|UNQ233/PRO266
- Immunogen:A synthetic peptide corresponding a region of human SLITRK1.
- Größe:50 µg
- VE:50 µG
Specifications
About this item
Members of the SLITRK family, such as SLITRK1, are integral membrane proteins with 2 N-terminal leucine-rich repeat (LRR) domains similar to those of SLIT proteins. Most SLITRKs, but not SLITRK1, also have C-terminal regions that share homology with neurotrophin receptors. SLITRKs are expressed predominantly in neural tissues and have neurite-modulating activity.Members of the SLITRK family, such as SLITRK1, are integral membrane proteins with 2 N-terminal leucine-rich repeat (LRR) domains similar to those of SLIT proteins (see SLIT1; MIM 603742). Most SLITRKs, but not SLITRK1, also have C-terminal regions that share homology with neurotrophin receptors (see NTRK1; MIM 191315). SLITRKs are expressed predominantly in neural tissues and have neurite-modulating activity (Aruga et al., 2003 [PubMed 14557068]).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLITRK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat