Specifications
- Antigen-Name:Anthrax Toxin Lethal Factor
- Antigen-Symbol:ATLF
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Bacteria
- Antigen-Synonyme:Anthrax lethal factor|Anthrax lethal toxin endopeptidase component|LF|Anthrax LF|LEF_BACAN.|bacillus anthracis lethal factor|Lef
- Lagerungspuffer:PBS containing 0.02% sodium azide
- Immunogen:Anthrax lethal factor antibody antibody was raised against a synthetic peptide corresponding to 16 amino acids near the carboxy terminus of the Anthrax lethal factor protein.
- Validierte Anwendungen:E
- Reinigung:Affinity chromatography purified via peptide column
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Anthrax Lethal Factor Antibody: Anthrax infection is initiated by the inhalation, ingestion, or cutaneous contact with Bacillus anthracis endospores. B. anthracis produces three polypeptides that comprise the anthrax toxin: protective antigen (PA), lethal factor (LF), and edema factor (EF). PA binds to two related proteins on the cell surface; these are termed tumor epithelial marker 8 (TEM8)/anthrax toxin receptor (ATR) and capillary morphogenesis protein 2 (CMG2), although it is still unclear which is physiologically relevant. Following PA binding to its receptor, PA is cleaved into two fragments by a furin-like protease. The bound fragment binds both LF and EF; the resulting complex is then endocytosed which allows the translocation of LF and EF into the cytoplasm. LF is the primary toxin of anthrax and functions as a highly specific protease that cleaves members of the mitogen-activated protein kinase kinase (MAPKK) family near their amino terminus, interfering with MAPK signaling and inducing apoptosis.
Type:
Antigen: ATLF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria