Drug and metabolite standards, desmethylclozapine (norclozapine), LoGiCal®
Catalog # PROOLGCFOR0480.03
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- VE:10 mg
- Normen:Reference standard
- CAS Nr.:6104-71-8
- Bezeichnung:Desmethylclozapine (Norclozapine)
- Formel:C₁₇H₁₇ClN₄
- Bezeichnung:Desmethylclozapine (Norclozapine)