Shoe covers, Millenium Grip
MILEOGNOIXS
- Schuhgröße:XS
- Anwendungsbereich Outfit:Labor,Lebensmittel-Bereich
- Geschlecht:Unisex
- Riskobereich:Regen (wasserdicht/wasserabweisend)
- Farbe:Schwarz
- Seq:01
- VE:1 PAIR
These waterproof overshoes are designed to keep shoes clean and avoid dirt from clogging. They are slip resistant on wet and oily floors.
- TPU outsole with high elasticity
- Abrasion and hydrocarbon resistant outsole
- Reusable and washable with warm soapy water
- Can be used with several types of shoes
Suitable for catering, food-processing industry, wet grounds like water, detergent, grease and oil.
Colour: Black
: NF EN 13287; F0, SRC