Organic reference standard, 9-hydroxyrisperidone
Catalog # PROOLGCFOR0491.03
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- VE:10 mg
- Normen:Reference standard
- CAS Nr.:144598-75-4
- Bezeichnung:9-Hydroxyrisperidone
- Formel:C₂₃H₂₇FN₄O₃
- Bezeichnung:9-Hydroxyrisperidone