Detergents, neodisher® EM, liquid detergents for special diswasher
148-0165
- Bezeichnung:neodisher® EM
- Verpackt:5 litre plastic container
- VE:5 L
Neodisher® EM is an additive for emulsification or dispersion as well as foam suppression in special dishwashers in the cosmetics industry, research, pharmaceutical industry and chemical industry.
- Combination with alkaline and acid cleaners, emulsifies or disperses undissolved impurities
- Emulsifies and/or disperses fats, oils, paraffins and solid matter in aqueous solutions
- It also has an anti foaming action