Mycophenolate Mofetil, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Catalog No:
- 1448956.
- 1448967.
- 1448978.
- 1448989.
- Bezeichnung:
- Mycophenolate Mofetil
- Mycophenolate Mofetil Related Compound A
- Mycophenolate Mofetil Related Compound B
- Mycophenolate Sodium
- Synonym(s):
- 2-Morpholinoethyl (E)-6-(1,3-dihydro-4,6-dihydroxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoate
- (RS)-7-hydroxy-5-methoxy-4-methyl-6-[2-(5-methyl-2-oxo-tetrahydrofuran-5-yl)ethyl]-3H-isobenzofuranyl-1-one
- CAS Nr.:
- 128794-94-5
- 1322681-36-6
- 26675-76-3
- 37415-62-6
- Formel:
- C₂₃H₃₁NO₇
- C₂₂H₂₉NO₇
- C₁₇H₂₀O₆
- C₁₇H₁₉NaO₆
- UN No.:
- 3077
Product Family Options
- BezeichnungNormenPackungVE
- Mycophenolate MofetilUSP reference standardVial200 mg1 * 200 mg406,00 €
Specifications
Cat. No.1448956.BezeichnungMycophenolate MofetilSynonym(s)CAS Nr.128794-94-5FormelC₂₃H₃₁NO₇UN No. - Mycophenolate Mofetil Related Compound AUSP reference standardVial15 mg1 * 15 mg1 300,00 €
Specifications
Cat. No.1448967.BezeichnungMycophenolate Mofetil Related Compound ASynonym(s)2-Morpholinoethyl (E)-6-(1,3-dihydro-4,6-dihydroxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoateCAS Nr.1322681-36-6FormelC₂₂H₂₉NO₇UN No. - Mycophenolate Mofetil Related Compound BUSP reference standardVial15 mg1 * 15 mg1 240,00 €
Specifications
Cat. No.1448978.BezeichnungMycophenolate Mofetil Related Compound BSynonym(s)(RS)-7-hydroxy-5-methoxy-4-methyl-6-[2-(5-methyl-2-oxo-tetrahydrofuran-5-yl)ethyl]-3H-isobenzofuranyl-1-oneCAS Nr.26675-76-3FormelC₁₇H₂₀O₆UN No. - Mycophenolate SodiumUSP reference standardVial200 mg1 * 200 mg382,00 €
Specifications
Cat. No.1448989.BezeichnungMycophenolate SodiumSynonym(s)CAS Nr.37415-62-6FormelC₁₇H₁₉NaO₆UN No.3077