Calcinated petroleum coke - trace elements
Catalog # PROONIST-2719
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- VE:50 g
- Bezeichnung:NIST-2719, calcined petroleum coke
Specifications
About this item
This reference standard is intended primarily for use in the calibration of apparatus and the evaluation of techniques employed in the analysis of calcined petroleum coke and other materials with a similar matrix.
Certifications: Traceable to NIST and SRM numbers.