Standard reference material, spinach leaves - trace elements, NIST-1570A
Catalog # PROONIST-1570A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- VE:60 g
- Bezeichnung:Spinach leaves - trace elements
Specifications
About this item
This standard material is intended for use in evaluating the reliability of analytical methods for the determination of major, minor, and trace elements, proximates, calories and total dietary fiber in botanical materials, agricultural food products, and materials of similar matrix.
A unit consists of 60 g of finely powdered dried spinach leaves.
Certifications: Traceable to NIST and SRM numbers.