AuftragsverfolgungKontakt
VWR®, Vortex Mixer
Anbieter:   VWR Collection
VWR®, Vortex Mixer
Anbieter:  VWR Collection
Zertifikate
VWR®, Vortex Mixer
2 Optionen unten verfügbar
Product Details & Documents
These vortex mixers are ideal for applications that demand repeatable results.
  • Cup head and three inch platform provided
  • LED display provided
  • Mixer speed: 3200 rpm
The control allows low rpm starts for gentle or high-speed mixing for vigorous vortexing of samples. The mixers have two modes of operation: Continuous mode when using optional accessory attachments or touch mode that activates mixing when the cup head is depressed. The microprocessor control maintains a set speed for strong, consistent mixing action. If users program the timer to a defined time limit, the unit shuts off when the time reaches zero. Displayed time counts up during continuous operation and counts down during timed runs.
 : All models include a cup head, a 76 mm head with cover and a 183 cm 3-wire cord and plug. Pulsing vortex mixer includes a stainless steel microtube holder with built in cup head and holds up to 1,5 to 2 ml microtubes.
Spezifikationen für diese Produktfamilie
Spezifikationen
  • Catalog No:
  • VWRU10153-842
  • VWRU10153-812
  • B×T×H:
  • 111,7×177,8×134,6 mm
  • 111,7×177,8×134,6 mm
  • Gewicht:
  • 4,5 kg
  • 4,5 kg
Spezifikationen
Product Family Options
Produktinformationen
  • Bezeichnung
    Verfügbarkeit
    Preis
  • Digital vortex mixer, 120 V
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
    Spezifikationen:
    Mehr
    Specifications
    Cat. No.
    VWRU10153-842
    B×T×H
    111,7×177,8×134,6 mm
    Gewicht
    4,5 kg
  • Pulsing vortex mixer, 120 V
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST - 1 190,00 €
    1 190,00 €
    1 * 1 ST
       1 190,00 €
    Spezifikationen:
    Mehr
    Specifications
    Cat. No.
    VWRU10153-812
    B×T×H
    111,7×177,8×134,6 mm
    Gewicht
    4,5 kg