ContactUsLinkComponent
Organic reference standards, Clomipramine, BP
Organic reference standards, Clomipramine, BP
Certificates
Organic reference standards, Clomipramine, BP
Specifications
  • CAS Nr.:
    32943-25-2
  • Formel:
    C₁₄H₁₂ClN
  • Bezeichnung:
    Clomipramine Impurity F