Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organische Standards
Organic reference standards, Cefradine assay standard, BP
Organic reference standards, Cefradine assay standard, BP
Organic reference standards, Cefradine assay standard, BP
PROOBP063
:
LGC Standards PROMOCHEM
:
Organic reference standards, Cefradine assay standard, BP
PROOBP063
:
LGC Standards PROMOCHEM
:
VE:
300 mg
Normen:
Reference standards
CAS Nr.:
38821-53-3
Bezeichnung:
Cefradine
Formel:
C₁₆H₁₉N₃O₄S
Bezeichnung:
Cefradine