ContactUsLinkComponent
Oxygen demand, quality control standard
Catalog # HACH2833510
Supplier:  Hach
CAS Number:  
undefined
Oxygen demand, quality control standard
Catalog # HACH2833510
Supplier:  Hach
CAS Number:  
Specifications
  • Konzentration:
    396 mg/l as BOD
    613 mg/l as COD
    242 mg/l as TOC
  • VE:
    1
  • Bezeichnung:
    Oxygen quality control standard, 10 ml Voluette® ampoules
Specifications
About this item
These mixed standards are ready to use, no dilutions are required. They contain BOD, COD and TOC.
Ordering information: Pack of 16×10 ml ampoules.