Test kit, photometrisch, Zinn, LCK-Küvetten-Test
Artikelnr. 705-3042
Lieferant: Hach
CAS-Nummer:
Spezifikationen
- Chemische Methode:Pyridinfluoron (PYF)
- VE:24 Tests
- Bezeichnung:Zinn
- Seq:278
- Typ:LCK359
- Für Gerät:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Messbereich:0,1 - 2,0 mg/l Sn
Spezifikationen
Über diesen Artikel
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Lagertemperatur: 2 - 8 °C (Kühlschranklagerung)