Disposable paper cups, PE
Artikelnr. 213-1098
Lieferant: NEOLAB
Spezifikationen
- Kapazität:200 ml
- Farbe:Weiß
- Höhe:95 mm
- Ext. Ø:70 mm
- VE:100
Spezifikationen
Über diesen Artikel
These paper cups feature a PE coating that prevents moisture penetration of the cup with moisture or humidity.
- Polyethylene is compostable
- Food safe
Ideal for food applications. Also suitable for filling, measuring, reviewing or mixing of the liquids or powders.