Order Entry
Austria
ContactUsLinkComponent
Organic reference standards, carvedilols, Ph. Eur.
Organic reference standards, carvedilols, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, carvedilols, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS Nr.:
    72956-09-3
  • Formel:
    C₂₄H₂₆N₂O₄
  • Bezeichnung:
    Carvedilol

Specifications

Frequently Bought Together